siRNA Details
Gene Symbol: CCDC88C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080414.3 | NP_001073883.2 | 2 | 282 | ||
XM_005267691.4 | XP_005267748.1 | 2 | 936 | |||
XM_011536796.2 | XP_011535098.1 | 2 | 236 | |||
XM_017021335.1 | XP_016876824.1 | 2 | 936 | |||
XM_017021337.1 | XP_016876826.1 | 2 | 936 | |||
GenBank mRNA | AF070587.1 | - | 2 | 246 | ||
AK302839.1 | - | 1 | 62 | |||
BC032316.1 | - | 2 | 254 | |||
DC402942.1 | - | 1 | 62 | |||
DR006516.1 | - | 2 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
