siRNA Details
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128215.1 | NP_001121687.1 | 9 | 1302 | ||
XM_011539748.2 | XP_011538050.1 | 9 | 1294 | |||
XM_011539751.2 | XP_011538053.1 | 8 | 4663 | |||
XM_011539752.2 | XP_011538054.1 | 7 | 663 | |||
GenBank mRNA | BC157888.1 | - | 10 | 1270 | 8862709 | |
BC171908.1 | - | 10 | 1270 | 9054562 | ||
EF426484.1 | - | 9 | 1292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
