siRNA Details
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128215.1 | NP_001121687.1 | 4 | 643 | ||
XM_011539748.2 | XP_011538050.1 | 4 | 614 | |||
XM_011539751.2 | XP_011538053.1 | 3 | 3983 | |||
XM_011539753.2 | XP_011538055.1 | 4 | 614 | |||
GenBank mRNA | BC157888.1 | - | 5 | 611 | 8862709 | |
BC171908.1 | - | 5 | 611 | 9054562 | ||
EF426484.1 | - | 4 | 633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
