siRNA Details
Gene Symbol: TMEM251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098621.1 | NP_001092091.1 | 2 | 1141 | ||
NM_015676.1 | NP_056491.1 | 2 | 767 | |||
GenBank mRNA | AL080118.1 | - | 2 | 736 | ||
BC030119.2 | - | 2 | 734 | |||
BP398505.1 | - | 1 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
