siRNA Details
Gene Symbol: IGHV1OR15-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135694.1 | 2 | 208 | |||
GenBank mRNA | HY100667.1 | - | 2 | 211 |
Gene Symbol: LOC102724760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135695.1 | 2 | 208 | |||
GenBank mRNA | DN998389.1 | - | 2 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
