siRNA Details
Gene Symbol: CC2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032449.2 | NP_115825.1 | 7 | 794 | ||
XM_005270590.2 | XP_005270647.1 | 7 | 880 | |||
XM_005270591.2 | XP_005270648.1 | 7 | 880 | |||
XM_005270593.2 | XP_005270650.1 | 3 | 663 | |||
XM_006710423.2 | XP_006710486.1 | 7 | 880 | |||
XM_017000559.1 | XP_016856048.1 | 7 | 810 | |||
GenBank mRNA | AB058739.1 | - | 3 | 225 | ||
BC014080.2 | - | 3 | 293 | |||
BC039308.1 | - | 6 | 1204 | |||
BQ073406.1 | - | 7 | 716 | |||
BX337298.2 | - | 7 | 781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
