siRNA Details
Gene Symbol: CC2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032449.2 | NP_115825.1 | 17, 18 | 2111 | ||
XM_005270590.2 | XP_005270647.1 | 17, 18 | 2197 | |||
XM_005270591.2 | XP_005270648.1 | 17, 18 | 2179 | |||
XM_005270593.2 | XP_005270650.1 | 13, 14 | 1933 | |||
XM_006710423.2 | XP_006710486.1 | 16, 17 | 2071 | |||
XM_017000559.1 | XP_016856048.1 | 17, 18 | 2109 | |||
GenBank mRNA | AB058739.1 | - | 13, 14 | 1542 | ||
BC007912.2 | - | 5, 6 | 559 | |||
BC014080.2 | - | 13, 14 | 1593 | |||
BC016880.1 | - | 11, 12 | 1302 | 3913149 | ||
BC039308.1 | - | 15, 16 | 2935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
