siRNA Details
Gene Symbol: DDX60L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012967.2 | NP_001012985.2 | 25 | 3479 | ||
NM_001291510.1 | NP_001278439.1 | 25 | 3479 | |||
XM_005263341.4 | XP_005263398.2 | 25 | 3462 | |||
XM_005263342.2 | XP_005263399.2 | 25 | 3479 | |||
XM_005263343.2 | XP_005263400.2 | 25 | 3445 | |||
XM_005263345.4 | XP_005263402.2 | 25 | 3458 | |||
XM_006714408.2 | XP_006714471.1 | 26 | 3529 | |||
XM_011532402.2 | XP_011530704.1 | 26 | 3752 | |||
XM_011532403.2 | XP_011530705.1 | 26 | 3587 | |||
XM_011532404.1 | XP_011530706.1 | 24 | 3383 | |||
XM_011532405.1 | XP_011530707.1 | 24 | 3383 | |||
XM_011532406.1 | XP_011530708.1 | 25 | 3479 | |||
XM_011532409.2 | XP_011530711.1 | 13 | 1519 | |||
XM_017008829.1 | XP_016864318.1 | 24 | 3349 | |||
XM_017008830.1 | XP_016864319.1 | 25 | 3479 | |||
XM_017008831.1 | XP_016864320.1 | 14 | 1781 | |||
GenBank mRNA | AK023743.1 | - | 4 | 364 | ||
AK055595.1 | - | 16 | 1978 | |||
AK092461.1 | - | 18 | 2319 | |||
AL832950.1 | - | 6 | 574 | |||
CR933601.1 | - | 25 | 3454 | |||
KR632542.1 | - | 23 | 3231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
