siRNA Details
Gene Symbol: CCDC173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085447.1 | NP_001078916.1 | 9 | 1793 | ||
XM_011510589.1 | XP_011508891.1 | 9 | 1681 | |||
XM_011510590.1 | XP_011508892.1 | 9 | 1714 | |||
GenBank mRNA | AK301851.1 | - | 9 | 1761 | ||
BC015980.2 | - | 3 | 741 | |||
BC065905.1 | - | 4 | 761 | |||
BC117445.1 | - | 9 | 1736 | 40125996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
