siRNA Details
Gene Symbol: FAM69A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006605.4 | NP_001006606.2 | 5 | 988 | ||
NM_001252269.1 | NP_001239198.1 | 4 | 853 | |||
NM_001252270.1 | NP_001239199.1 | 4 | 811 | |||
NM_001252271.1 | NP_001239200.1 | 5 | 1031 | |||
GenBank mRNA | AK027146.1 | - | 2 | 549 | ||
AK303700.1 | - | 4 | 1006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
