siRNA Details
Gene Symbol: GOLGA7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010917.2 | NP_001010917.1 | 3 | 247 | ||
GenBank mRNA | BC110810.1 | - | 3 | 247 | 5195678 | |
DN831741.1 | - | 2 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
