siRNA Details
Gene Symbol: WHAMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080435.2 | NP_001073904.1 | 9 | 1868 | ||
XM_005272421.4 | XP_005272478.1 | 9 | 1484 | |||
XM_005272422.3 | XP_005272479.1 | 9 | 1231 | |||
XM_005272423.4 | XP_005272480.1 | 9 | 1470 | |||
XM_011521232.2 | XP_011519534.1 | 7 | 1680 | |||
GenBank mRNA | AB075851.1 | - | 9 | 1854 | ||
AK097947.1 | - | 1 | 70 | |||
AK126887.1 | - | 9 | 1870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
