siRNA Details
Gene Symbol: NAA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032693.2 | NP_116082.1 | 1 | 540 | ||
XM_017008777.1 | XP_016864266.1 | 1 | 581 | |||
XM_017008778.1 | XP_016864267.1 | 1 | 581 | |||
XM_017008779.1 | XP_016864268.1 | 1 | 581 | |||
GenBank mRNA | BC004552.2 | - | 1 | 540 | ||
BC063623.1 | - | 1 | 523 | |||
BC080651.1 | - | 1 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
