siRNA Details
Gene Symbol: SMTNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105565.2 | NP_001099035.2 | 5, 6 | 1187 | ||
XM_005273819.3 | XP_005273876.1 | 6, 7 | 1989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
