siRNA Details
Gene Symbol: FRRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013660.2 | NP_001013682.2 | 9 | 1567 | ||
XM_005270861.3 | XP_005270918.1 | 9 | 1094 | |||
XM_011541451.2 | XP_011539753.1 | 9 | 1567 | |||
XM_011541452.1 | XP_011539754.1 | 9 | 1094 | |||
XM_017001271.1 | XP_016856760.1 | 7 | 978 | |||
GenBank mRNA | AK131302.1 | - | 9 | 1567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
