siRNA Details
Gene Symbol: TATDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014760.3 | NP_055575.3 | 4 | 1860 | ||
GenBank mRNA | AK289794.1 | - | 4 | 1581 | ||
BC090935.1 | - | 4 | 1441 | 3064714 | ||
BC101770.1 | - | 4 | 1349 | 8069276 | ||
BC101776.1 | - | 4 | 1350 | 8069282 | ||
D86972.1 | - | 5 | 1677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
