siRNA Details
Gene Symbol: TATDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014760.3 | NP_055575.3 | 5 | 2500 | ||
GenBank mRNA | AK289794.1 | - | 5 | 2221 | ||
BC090935.1 | - | 5 | 2081 | 3064714 | ||
BC101770.1 | - | 5 | 1989 | 8069276 | ||
BC101776.1 | - | 5 | 1990 | 8069282 | ||
D86972.1 | - | 6 | 2317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
