siRNA Details
Gene Symbol: GHRLOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU789547.1 | - | 1 | 748 | ||
EU789548.1 | - | 1 | 748 | |||
EU789549.1 | - | 2 | 563 | |||
EU789550.1 | - | 1 | 748 |
Gene Symbol: TATDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014760.3 | NP_055575.3 | 8 | 3834 | ||
GenBank mRNA | BC090935.1 | - | 8 | 3415 | 3064714 | |
BG674405.1 | - | 1 | 289 | |||
D86972.1 | - | 9 | 3622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
