siRNA Details
Gene Symbol: UBN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173569.3 | NP_775840.3 | 9 | 1660 | ||
XM_005250249.3 | XP_005250306.2 | 9 | 3577 | |||
XM_006715916.3 | XP_006715979.2 | 8 | 3478 | |||
XM_011516002.2 | XP_011514304.1 | 9 | 3580 | |||
XM_011516003.1 | XP_011514305.1 | 9 | 3580 | |||
GenBank mRNA | AB107352.1 | - | 4 | 688 | ||
AK098644.1 | - | 3 | 230 | |||
AK125956.1 | - | 9 | 1440 | |||
BC037413.1 | - | 1 | 87 | |||
BC065034.1 | - | 6 | 966 | |||
BC108249.1 | - | 4 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
