siRNA Details
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168241.1 | NP_001161713.1 | 4 | 993 | ||
NM_001191033.1 | NP_001177962.1 | 3 | 2388 | |||
XM_006711951.3 | XP_006712014.1 | 4 | 949 | |||
XM_011532564.2 | XP_011530866.1 | 4 | 1430 | |||
XM_011532565.2 | XP_011530867.1 | 5 | 1003 | |||
XM_011532566.2 | XP_011530868.1 | 4 | 761 | |||
XM_011532567.2 | XP_011530869.1 | 4 | 956 | |||
GenBank mRNA | AB124552.1 | - | 4 | 993 | ||
AK090454.1 | - | 3 | 2390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
