siRNA Details
Gene Symbol: CIB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029881.2 | NP_001025052.1 | 6, 7 | 589 | ||
XM_011532514.2 | XP_011530816.1 | 6, 7 | 672 | |||
XM_011532517.2 | XP_011530819.1 | 4, 5 | 959 | |||
XM_017003329.1 | XP_016858818.1 | 5, 6 | 1443 | |||
XM_017003330.1 | XP_016858819.1 | 4, 5 | 1353 | |||
GenBank mRNA | BC127684.1 | - | 6, 7 | 589 | 40127392 | |
BC127685.1 | - | 6, 7 | 587 | 40127393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
