siRNA Details
Gene Symbol: CIB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029881.2 | NP_001025052.1 | 1 | 79 | ||
XM_011532514.2 | XP_011530816.1 | 1 | 106 | |||
XM_017003331.1 | XP_016858820.1 | 1 | 509 | |||
GenBank mRNA | BC127684.1 | - | 1 | 79 | 40127392 | |
BC127685.1 | - | 1 | 77 | 40127393 | ||
BC140923.1 | - | 1 | 53 | 9056735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
