siRNA Details
Gene Symbol: AAED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153698.1 | NP_714542.1 | 5, 6 | 586 | ||
XM_005251784.4 | XP_005251841.1 | 5, 6 | 438 | |||
GenBank mRNA | BC136501.1 | - | 5, 6 | 424 | 9020490 | |
BC136503.1 | - | 5, 6 | 586 | 9020492 | ||
BF445089.1 | - | 5, 6 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
