siRNA Details
Gene Symbol: SHROOM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020717.3 | NP_065768.2 | 4 | 2555 | ||
NR_027121.1 | 4 | 2555 | ||||
XM_017029682.1 | XP_016885171.1 | 4 | 2661 | |||
XM_017029683.1 | XP_016885172.1 | 3 | 2526 | |||
XM_017029684.1 | XP_016885173.1 | 3 | 2678 | |||
XM_017029685.1 | XP_016885174.1 | 4 | 2678 | |||
XM_017029686.1 | XP_016885175.1 | 2 | 2543 | |||
XM_017029687.1 | XP_016885176.1 | 4 | 2665 | |||
GenBank mRNA | AB033028.1 | - | 4 | 2555 | ||
BC151240.1 | - | 4 | 2566 | 8860513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
