siRNA Details
Gene Symbol: SHROOM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020717.3 | NP_065768.2 | 7 | 3846 | ||
NR_027121.1 | 7 | 3846 | ||||
XM_017029682.1 | XP_016885171.1 | 8 | 4066 | |||
XM_017029683.1 | XP_016885172.1 | 7 | 3931 | |||
XM_017029684.1 | XP_016885173.1 | 7 | 4083 | |||
XM_017029685.1 | XP_016885174.1 | 8 | 4083 | |||
XM_017029686.1 | XP_016885175.1 | 6 | 3948 | |||
GenBank mRNA | AB033028.1 | - | 7 | 3861 | ||
BC151240.1 | - | 7 | 3872 | 8860513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
