siRNA Details
Gene Symbol: ZSCAN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012455.1 | NP_001012458.1 | 4 | 1580 | ||
GenBank mRNA | AL833347.1 | - | 4 | 1545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
