siRNA Details
Gene Symbol: ZSCAN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012455.1 | NP_001012458.1 | 3 | 699 | ||
XM_005248950.2 | XP_005249007.1 | 3 | 697 | |||
XM_011514406.2 | XP_011512708.1 | 3 | 697 | |||
XM_011514408.2 | XP_011512710.1 | 4 | 884 | |||
XM_011514410.2 | XP_011512712.1 | 3 | 697 | |||
XM_017010479.1 | XP_016865968.1 | 3 | 3913 | |||
GenBank mRNA | AL833347.1 | - | 3 | 664 | ||
BC101557.1 | - | 2 | 601 | 8069063 | ||
BC101559.1 | - | 2 | 601 | 8069065 | ||
BX283350.1 | - | 2 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
