siRNA Details
Gene Symbol: SLC45A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080397.2 | NP_001073866.2 | 7 | 1819 | ||
XM_011541530.1 | XP_011539832.1 | 7 | 1820 | |||
XM_011541531.1 | XP_011539833.1 | 6 | 1727 | |||
GenBank mRNA | AF118274.1 | - | 3 | 1048 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
