siRNA Details
Gene Symbol: NPY6R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002713.3 | 2 | 745 | |||
GenBank mRNA | AK297855.1 | - | 1 | 253 | ||
AK299427.1 | - | 2 | 612 | |||
D86519.1 | - | 1 | 736 | |||
U59431.1 | - | 1 | 459 | |||
U67780.1 | - | 1 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
