siRNA Details
Gene Symbol: LOC440461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027283.1 | 1 | 1154 | |||
GenBank mRNA | BC080624.1 | - | 1 | 978 |
Gene Symbol: ARHGAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174919.3 | NP_777579.2 | 4 | 1353 | ||
GenBank mRNA | AF258593.1 | - | 5 | 2295 | ||
AK290650.1 | - | 4 | 1304 | |||
BC067345.1 | - | 4 | 1353 | 6084901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
