siRNA Details
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020759.2 | NP_065810.2 | 23 | 10035 | ||
XM_011521831.2 | XP_011520133.1 | 23 | 10048 | |||
XM_011521832.2 | XP_011520134.1 | 23 | 10045 | |||
XM_011521833.2 | XP_011520135.1 | 22 | 10015 | |||
XM_011521834.2 | XP_011520136.1 | 22 | 9985 | |||
XM_011521835.2 | XP_011520137.1 | 21 | 9907 | |||
XM_011521836.2 | XP_011520138.1 | 23 | 10350 | |||
XM_011521837.2 | XP_011520139.1 | 21 | 9817 | |||
XM_011521839.2 | XP_011520141.1 | 10 | 8817 | |||
XM_017022439.1 | XP_016877928.1 | 22 | 9970 | |||
XM_017022440.1 | XP_016877929.1 | 22 | 9947 | |||
XM_017022441.1 | XP_016877930.1 | 21 | 10266 | |||
XM_017022442.1 | XP_016877931.1 | 16 | 9374 | |||
XM_017022443.1 | XP_016877932.1 | 15 | 9358 | |||
GenBank mRNA | AB037721.1 | - | 1 | 1339 | ||
CR749416.1 | - | 1 | 4057 | |||
CR936665.1 | - | 1 | 1962 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
