siRNA Details
Gene Symbol: PRR29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164257.1 | NP_001157729.1 | 4 | 355 | ||
NM_001191029.1 | NP_001177958.1 | 4 | 355 | |||
NM_001191030.1 | NP_001177959.1 | 4 | 334 | |||
XM_005257805.4 | XP_005257862.1 | 4 | 355 | |||
XM_011525469.1 | XP_011523771.1 | 3 | 273 | |||
GenBank mRNA | AK021786.1 | - | 4 | 355 | ||
AK294360.1 | - | 3 | 279 | |||
AK302960.1 | - | 4 | 355 | |||
AK304254.1 | - | 4 | 334 | |||
BC030709.1 | - | 3 | 545 | |||
DC358808.1 | - | 4 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
