siRNA Details
Gene Symbol: PRR29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164257.1 | NP_001157729.1 | 6 | 571 | ||
NM_001191029.1 | NP_001177958.1 | 5 | 498 | |||
NM_001191030.1 | NP_001177959.1 | 6 | 550 | |||
NM_001191031.1 | NP_001177960.1 | 5 | 344 | |||
XM_005257805.4 | XP_005257862.1 | 5 | 766 | |||
XM_011525468.2 | XP_011523770.1 | 3 | 420 | |||
XM_011525469.1 | XP_011523771.1 | 5 | 489 | |||
XM_017025315.1 | XP_016880804.1 | 4 | 225 | |||
GenBank mRNA | AK021786.1 | - | 6 | 571 | ||
AK294360.1 | - | 5 | 495 | |||
AK302960.1 | - | 5 | 498 | |||
AK304254.1 | - | 6 | 550 | |||
AL161972.1 | - | 2 | 68 | |||
BC030709.1 | - | 5 | 761 | |||
BC045676.1 | - | 4 | 523 | |||
DB037384.1 | - | 5 | 344 | |||
DC358808.1 | - | 6 | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
