siRNA Details
Gene Symbol: C16orf86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012984.2 | NP_001013002.2 | 4 | 1029 | ||
XM_005255952.4 | XP_005256009.1 | 4 | 1064 | |||
GenBank mRNA | AK123796.1 | - | 2 | 1480 | ||
BC064498.1 | - | 4 | 1075 | |||
BC082249.1 | - | 4 | 1075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
