siRNA Details
Gene Symbol: FAM205BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024481.1 | 4 | 4117 | |||
GenBank mRNA | AL080137.1 | - | 1 | 1368 | ||
BX648118.1 | - | 4 | 4063 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
