siRNA Details
Gene Symbol: ZNF229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278510.2 | NP_001265439.2 | 6 | 756 | ||
NM_014518.3 | NP_055333.3 | 6 | 885 | |||
NR_103551.2 | 6 | 1340 | ||||
XM_006723372.3 | XP_006723435.1 | 6 | 959 | |||
XM_011527292.1 | XP_011525594.1 | 6 | 867 | |||
XM_017027280.1 | XP_016882769.1 | 6 | 1414 | |||
GenBank mRNA | AB209357.1 | - | 6 | 1309 | ||
AF192979.1 | - | 6 | 886 | |||
AY166791.1 | - | 2 | 292 | |||
BC150612.1 | - | 4 | 621 | 9056982 | ||
CR936724.1 | - | 6 | 757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
