siRNA Details
Gene Symbol: ZNF229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278510.2 | NP_001265439.2 | 6 | 920 | ||
NM_014518.3 | NP_055333.3 | 6 | 1049 | |||
NR_103551.2 | 6 | 1504 | ||||
XM_006723372.3 | XP_006723435.1 | 6 | 1123 | |||
XM_011527292.1 | XP_011525594.1 | 6 | 1031 | |||
XM_017027280.1 | XP_016882769.1 | 6 | 1578 | |||
GenBank mRNA | AB209357.1 | - | 6 | 1473 | ||
AF192979.1 | - | 6 | 1050 | |||
BC150612.1 | - | 4 | 785 | 9056982 | ||
CR936724.1 | - | 6 | 921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
