siRNA Details
Gene Symbol: CCDC58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017928.3 | NP_001017928.1 | 5 | 507 | ||
NM_001308326.1 | NP_001295255.1 | 5 | 533 | |||
XM_011512410.2 | XP_011510712.1 | 5 | 485 | |||
GenBank mRNA | BC054051.1 | - | 5 | 488 | ||
BC062725.1 | - | 5 | 494 | 6495960 | ||
BC109378.1 | - | 5 | 481 | |||
CD683826.1 | - | 5 | 531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
