siRNA Details
Gene Symbol: ZNF714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182515.3 | NP_872321.2 | 5 | 2382 | ||
NR_117086.1 | 6 | 2401 | ||||
NR_117087.1 | 5 | 2382 | ||||
GenBank mRNA | BC022527.1 | - | 6 | 2343 | ||
BC037782.1 | - | 6 | 2348 | 4797729 | ||
BC050468.2 | - | 5 | 2320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
