siRNA Details
Gene Symbol: PHLDB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198850.3 | NP_942147.3 | 16 | 2278 | ||
XM_005259169.3 | XP_005259226.1 | 16 | 2346 | |||
XM_005259170.3 | XP_005259227.1 | 16 | 2244 | |||
XM_011527236.2 | XP_011525538.1 | 16 | 2450 | |||
GenBank mRNA | BC007947.2 | - | 5 | 633 | ||
BC157852.1 | - | 10 | 1160 | |||
BC171853.1 | - | 10 | 1169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
