siRNA Details
Gene Symbol: GATS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178831.6 | NP_849153.3 | 5 | 1954 | ||
NR_028039.1 | 8 | 2766 | ||||
NR_028040.1 | 6 | 2078 | ||||
XM_017012154.1 | XP_016867643.1 | 6 | 1980 | |||
XM_017012155.1 | XP_016867644.1 | 6 | 1956 | |||
XM_017012157.1 | XP_016867646.1 | 5 | 1869 | |||
GenBank mRNA | AK056608.1 | - | 5 | 1460 | ||
AK095056.1 | - | 4 | 1468 | |||
BC033723.1 | - | 5 | 1947 | |||
BC065200.1 | - | 7 | 2083 | |||
BC090867.1 | - | 8 | 2741 | 6179098 | ||
CR749506.1 | - | 6 | 1829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
