siRNA Details
Gene Symbol: ZNF251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138367.1 | NP_612376.1 | 4 | 480 | ||
XM_005272356.4 | XP_005272413.1 | 4 | 498 | |||
XM_006716683.2 | XP_006716746.1 | 3 | 299 | |||
XM_011517361.1 | XP_011515663.1 | 3 | 299 | |||
GenBank mRNA | AK000435.1 | - | 4 | 364 | ||
AK091638.1 | - | 2 | 2287 | |||
DA811573.1 | - | 4 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
