siRNA Details
Gene Symbol: ZNF251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138367.1 | NP_612376.1 | 5 | 1804 | ||
GenBank mRNA | AK000435.1 | - | 5 | 1688 | ||
AK027286.1 | - | 1 | 840 | |||
BC006258.2 | - | 1 | 415 | |||
BC093984.1 | - | 1 | 324 | |||
BC112137.1 | - | 1 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
