siRNA Details
Gene Symbol: YDJC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017964.1 | NP_001017964.1 | 5 | 1176 | ||
XM_011529906.2 | XP_011528208.1 | 4 | 1273 | |||
GenBank mRNA | BC096754.1 | - | 5 | 1249 | ||
BC110075.1 | - | 4 | 982 | |||
BC110799.1 | - | 5 | 1220 | |||
BC133043.1 | - | 5 | 1330 | |||
BC133047.1 | - | 5 | 1330 | |||
BQ050386.1 | - | 1 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
