siRNA Details
Gene Symbol: C19orf81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195076.1 | NP_001182005.1 | 4 | 278 | ||
XM_011526910.2 | XP_011525212.1 | 4 | 468 | |||
GenBank mRNA | BX281890.1 | - | 3 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
