siRNA Details
Gene Symbol: FAM171A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198475.2 | NP_940877.2 | 6 | 961 | ||
XM_017024490.1 | XP_016879979.1 | 4 | 528 | |||
GenBank mRNA | BC029298.1 | - | 6 | 977 | ||
BC030200.1 | - | 6 | 961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
