siRNA Details
Gene Symbol: SYCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105578.1 | NP_001099048.1 | 6 | 693 | ||
XM_005259848.4 | XP_005259905.1 | 6 | 811 | |||
XM_011527882.2 | XP_011526184.1 | 6 | 1092 | |||
XM_011527883.2 | XP_011526185.1 | 6 | 1138 | |||
GenBank mRNA | AK097443.1 | - | 5 | 1989 | ||
AK302373.1 | - | 6 | 683 | |||
BC035819.2 | - | 5 | 662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
