siRNA Details
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199107.1 | NP_001186036.1 | 2 | 85 | ||
NM_020705.2 | NP_065756.1 | 2 | 85 | |||
XM_017023493.1 | XP_016878982.1 | 2 | 127 | |||
XM_017023494.1 | XP_016878983.1 | 2 | 215 | |||
XM_017023495.1 | XP_016878984.1 | 2 | 127 | |||
GenBank mRNA | AB032997.1 | - | 2 | 78 | ||
BC112389.1 | - | 2 | 85 | 5502314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
