siRNA Details
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199107.1 | NP_001186036.1 | 8 | 4076 | ||
NM_020705.2 | NP_065756.1 | 7 | 4058 | |||
XM_017023493.1 | XP_016878982.1 | 9 | 3381 | |||
XM_017023494.1 | XP_016878983.1 | 7 | 4188 | |||
XM_017023495.1 | XP_016878984.1 | 8 | 3363 | |||
GenBank mRNA | AB032997.1 | - | 8 | 4069 | ||
AK074509.1 | - | 1 | 1223 | |||
BC112389.1 | - | 7 | 4058 | 5502314 | ||
BX648283.1 | - | 4 | 2805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
