siRNA Details
Gene Symbol: BICDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255135.4 | XP_005255192.1 | 10 | 1952 | ||
XM_011522391.1 | XP_011520693.1 | 9 | 2140 | |||
GenBank mRNA | AL833717.1 | - | 7 | 3097 | ||
AL833749.1 | - | 8 | 3416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
